Learn More
Cayman Chemical Ospemifene-d4, Purity: ≥99% deuterated forms (d1-d4), 1 mg
SDP
Catalog No. 502949931
Encompass_Preferred
An internal standard for the quantification of ospemifene by GC- or LC-MS
Catalog No. 50-294-9931 Supplier Cayman Chemical Supplier No. 334651 MG
An internal standard for the quantification of ospemifene by GC- or LC-MS