Learn More
Cayman Chemical OspemIfen-d4 5mg
SDP
Catalog No. 502949932
Encompass_Preferred
An internal standard for the quantification of ospemifene by GC- or LC-MS
Catalog No. 50-294-9932 Supplier Cayman Chemical Supplier No. 334655 MG
An internal standard for the quantification of ospemifene by GC- or LC-MS