Learn More
Medchemexpress LLC Ospemifene-d4 | 128607-22-7 | 99.7% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0723S1MG
Encompass_Preferred
Ospemifene-d4 is a deuterium-labeled Ospemifene. Ospemifene is a selective and orally active estrogen receptor modulator used for the prevention of osteoporosis. It has IC50 values of 827 nM for estrogen receptor α (ERα) and 1633 nM for ERβ. Additionally, Ospemifene exhibits bone-sparing, antitumor, and cholesterol-lowering effects.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-05256