Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ospemifene 98.0+%, TCI America™
SDP

Catalog No. O04411G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04411G 1 g
O0441100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O04411G Supplier TCI America Supplier No. O04411G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 128607-22-7
Molecular Formula C24H23ClO2
Molecular Weight (g/mol) 378.896
MDL Number MFCD00871890
InChI Key LUMKNAVTFCDUIE-VHXPQNKSSA-N
PubChem CID 3036505
ChEBI CHEBI:73275
IUPAC Name 2-[4-[(Z)-4-chloro-1,2-diphenylbut-1-enyl]phenoxy]ethanol
SMILES C1=CC=C(C=C1)C(=C(C2=CC=CC=C2)C3=CC=C(C=C3)OCCO)CCCl

Specifications

Color White
Quantity 1 g
Formula Weight 378.90
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ospemifene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.