Learn More
Bulbconnection Dot Com OSRAM 54261 64250 HLX 6V20W
Catalog No. NC1000906
Encompass
This Osram Lamp Has An Ansi Code Of Esb. The Bulb Type Of This Osram 64250 Hlx Bulb Is Type T-3. The Base Type Of The Osram 64250 Hlx Replacement Lamp May Also Be Called T3. The Osram 64250 Hlx Replacement Lamp Has A Base Type Of G4 (Miniature 2-Pin). This Osram 64250 Hlx Lamp Is A 20 Watt Bulb. The Osram 64250 Hlx Lamp Is A 6 Volt Bulb. The Maximum Overall Length Of The Osram 64250 Hlx Replacement Lamp Is 1.209 Inches Or 30.7 Millimeters. The Bulb Diameter Of The Osram 64250 Hlx Replacement Bulb Is 0.354 Inches Or 9.0 Millimeters. The Light Center Length Of This Osram 64250 Hlx Bulb Is 0.761 Inches Or 19.3 Millimeters.
Catalog No. NC1000906 Supplier Bulbconnection Dot Com Supplier No. BC1514
This Osram Lamp Has An Ansi Code Of Esb. The Bulb Type Of This Osram 64250 Hlx Bulb Is Type T-3. The Base Type Of The Osram 64250 Hlx Replacement Lamp May Also Be Called T3. The Osram 64250 Hlx Replacement Lamp Has A Base Type Of G4 (Miniature 2-Pin). This Osram 64250 Hlx Lamp Is A 20 Watt Bulb. The Osram 64250 Hlx Lamp Is A 6 Volt Bulb. The Maximum Overall Length Of The Osram 64250 Hlx Replacement Lamp Is 1.209 Inches Or 30.7 Millimeters. The Bulb Diameter Of The Osram 64250 Hlx Replacement Bulb Is 0.354 Inches Or 9.0 Millimeters. The Light Center Length Of This Osram 64250 Hlx Bulb Is 0.761 Inches Or 19.3 Millimeters.