Learn More
Bulbman Inc 5761 - 64265 HLX Osram Photo Optic 54606 Light Bulb: 30 watt, 6 volt, clear T3.5 quartz halogen lamp with a G4 bipin base
SDP
Supplier:  Bulbman Inc 136980
Encompass_Preferred
5761 - 64265 HLX Osram Photo Optic 54606 Light Bulb: 30 watt, 6 volt, clear T3.5 quartz halogen lamp with a G4 bipin base.WARM WHITE (2600-3200 KELVIN)Precise filament alignment with patented quartz pinch technologyOSRAM HLX lamps provide 10% higher luminous efficacy over standard halogenInstant on and nearly constant luminous flux over the life of the lampDimmable
Catalog No. NC1619610