Learn More
Bulbman Inc Osram BAB/CC, Sylvania Tru Aim Brilliant Tungsten Halogen Bulb, 20 Watt, 12 Volt
SDP
Catalog No. 501253615
Encompass_Preferred
Osram BAB/CC: Sylvania Tru Aim Brilliant Tungsten Halogen MR16 Brilliant Axial Filament UV Filter 20MR16/B/FL35 12 volt, 20 watt, 35 degree flood. Sylvania TRU-AIM BRILLIANT MR16 lamps have an aluminized reflector that does not deteriorate over time. Therefore, they continue to have highly stable color and crisp, white light output throughout their service life, consistent color and transmits heat through the front of the lamp to reduce fixture damage
Catalog No. 50-125-3615 Supplier Bulbman Inc Supplier No. 130849
Osram BAB/CC: Sylvania Tru Aim Brilliant Tungsten Halogen MR16 Brilliant Axial Filament UV Filter 20MR16/B/FL35 12 volt, 20 watt, 35 degree flood. Sylvania TRU-AIM BRILLIANT MR16 lamps have an aluminized reflector that does not deteriorate over time. Therefore, they continue to have highly stable color and crisp, white light output throughout their service life, consistent color and transmits heat through the front of the lamp to reduce fixture damage