Learn More
Bulbman Inc OSRAM EFR 15 Volt, 150 Watt Halogen Light Bulb
SDP
Catalog No. NC1129226
Encompass_Preferred
15 volt, 150 watt Xenophot reflector EFR (64634 HLX) bulb applications: medical and dental equipment projectors microscopes fiber optic illuminator.
Catalog No. NC1129226 Supplier Bulbman Inc Supplier No. 103240
15 volt, 150 watt Xenophot reflector EFR (64634 HLX) bulb applications: medical and dental equipment projectors microscopes fiber optic illuminator.