Learn More
Bulbman Inc Osram ENX 54984 Tungsten Halogen Bulb, 360 Watt, 82 Volt
SDP
Catalog No. 501253547
Encompass_Preferred
Osram ENX: The Osram ENX is a 82 volt (82V) 360 watt (360W), tungsten halogen, low to medium voltage with reflector. Typical lifetime is 75 hours. This lamp has a MR16 reflector with GX5.3 bi-pin base. This item is made by Osram, but has alternative part numbers: 54984 and ENX 82V-360W.
Catalog No. 50-125-3547 Supplier Bulbman Inc Supplier No. 103539
Osram ENX: The Osram ENX is a 82 volt (82V) 360 watt (360W), tungsten halogen, low to medium voltage with reflector. Typical lifetime is 75 hours. This lamp has a MR16 reflector with GX5.3 bi-pin base. This item is made by Osram, but has alternative part numbers: 54984 and ENX 82V-360W.