Learn More
Bulbman Inc XBO 75W/2 Osram 69231
SDP
Catalog No. 501253544
Encompass_Preferred
Osram XBO75W/2 69231 : The Osram XBO 75 W/2 is a 75 watt, ozone generating quartz Xenon Short Arc Lamp with 1000 lumens visible light output. Typical lifetime is 400 hours. This lamp has SFa7.5-2 (anode) and SFa7.5-2 (cathode) basing on the 10mm diameter x 90mm long clear double ended bulb. This item is made by Osram, and is commonly available from no other source but Osram/Sylvania, but as alternative part numbers 69231, XBO75W/2, XBO75W/2, XBO75W/2, Olympus 8-B198, Nikon 78682, Zeiss 380079-9190 and other variations.
Catalog No. 50-125-3544 Supplier Bulbman Inc Supplier No. 106226
Osram XBO75W/2 69231 : The Osram XBO 75 W/2 is a 75 watt, ozone generating quartz Xenon Short Arc Lamp with 1000 lumens visible light output. Typical lifetime is 400 hours. This lamp has SFa7.5-2 (anode) and SFa7.5-2 (cathode) basing on the 10mm diameter x 90mm long clear double ended bulb. This item is made by Osram, and is commonly available from no other source but Osram/Sylvania, but as alternative part numbers 69231, XBO75W/2, XBO75W/2, XBO75W/2, Olympus 8-B198, Nikon 78682, Zeiss 380079-9190 and other variations.