Learn More
Bulbman Inc OSRAM XBOR100W/45-OFR 69197
SDP
Catalog No. 501253540
Encompass_Preferred
Osram XBOR100W/45-OFR 69197: The Osram XBO R 100 W/45 OFR is a 100 watt, ozone free quartz Xenon Short Arc Lamp with focussing 44 degree beam angle. Typical lifetime is 500 hours. This lamp has contact pins front and back on the 67mm diameter x 83mm long dichroic coated elliptical reflector with a 45mm working distance. This item is made by Osram, and is commonly available from no other source but Osram/Sylvania, but as alternative part numbers 69197, XBOR100W45, XBO-R100W/45, XBOR100W/45, XBO-R100W/45 OFR, XBOR100W/45 OFR and other variations.
Catalog No. 50-125-3540 Supplier Bulbman Inc Supplier No. 108873
Osram XBOR100W/45-OFR 69197: The Osram XBO R 100 W/45 OFR is a 100 watt, ozone free quartz Xenon Short Arc Lamp with focussing 44 degree beam angle. Typical lifetime is 500 hours. This lamp has contact pins front and back on the 67mm diameter x 83mm long dichroic coated elliptical reflector with a 45mm working distance. This item is made by Osram, and is commonly available from no other source but Osram/Sylvania, but as alternative part numbers 69197, XBOR100W45, XBO-R100W/45, XBOR100W/45, XBO-R100W/45 OFR, XBOR100W/45 OFR and other variations.