Learn More
Selleck Chemical LLC OSS_128167, 10mM (1mL in DMSO)
SDP
Catalog No. NC2937804
Encompass_Preferred
OSS_128167 (SIRT6-IN-1) is a specific SIRT6 inhibitor with IC50 values of 89, 1578 and 751 μM for SIRT6, SIRT1 and SIRT2, respectively. OSS_128167 has anti-viral effect in HBV transcription and replication.
Catalog No. NC2937804 Supplier Selleck Chemical LLC Supplier No. S862710MM/1ML
OSS_128167 (SIRT6-IN-1) is a specific SIRT6 inhibitor with IC50 values of 89, 1578 and 751 μM for SIRT6, SIRT1 and SIRT2, respectively. OSS_128167 has anti-viral effect in HBV transcription and replication.