Learn More
Selleck Chemical LLC OSS_128167 5mg 887686-02-4 SIRT6-IN-1
SDP
Supplier:  Selleck Chemical LLC S86275MG
Encompass_Preferred
OSS_128167 (SIRT6-IN-1) is a specific SIRT6 inhibitor with IC50 values of 89, 1578 and 751 ?M for SIRT6, SIRT1 and SIRT2, respectively. OSS_128167 has anti-viral effect in HBV transcription and replication. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8050