Learn More
Medchemexpress LLC Oss 128167 | 887686-02-4 | 98.6% | 366.32 | C19H14N2O6 | 25 MG
SDP
Catalog No. 5000239874
Encompass_Preferred
OSS_128167 is a research-use small molecule that selectively inhibits sirtuin 6 (SIRT6). It has been characterized in cellular and animal studies and shows activity on histone acetylation, metabolic regulation, antiviral effects against hepatitis B virus, and chemosensitization in cancer models. Supplied as a high-purity solid.
  • Selective SIRT6 inhibition (IC50 = 89 μM).
  • Lower activity against SIRT1 and SIRT2 (IC50s = 1578 μM and 751 μM).
  • Demonstrated anti-HBV activity in vitro and in vivo.
  • Increases H3K9 acetylation and GLUT-1 expression in cells.
  • Induces chemosensitization in multiple myeloma cell lines.
  • High reported purity and supplied as a white to light yellow solid.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-39874 Supplier Medchemexpress LLC Supplier No. HY10745425MG
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
OSS_128167 is a research-use small molecule that selectively inhibits sirtuin 6 (SIRT6). It has been characterized in cellular and animal studies and shows activity on histone acetylation, metabolic regulation, antiviral effects against hepatitis B virus, and chemosensitization in cancer models. Supplied as a high-purity solid.
  • Selective SIRT6 inhibition (IC50 = 89 μM).
  • Lower activity against SIRT1 and SIRT2 (IC50s = 1578 μM and 751 μM).
  • Demonstrated anti-HBV activity in vitro and in vivo.
  • Increases H3K9 acetylation and GLUT-1 expression in cells.
  • Induces chemosensitization in multiple myeloma cell lines.
  • High reported purity and supplied as a white to light yellow solid.
  • For research use only; not for human or clinical use.