Learn More
Aviva Systems Biology Corp OSTalpha Antibody - middle region (ARP53102_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53102_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OSTalpha. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human OSTalpha. Alternate names: MGC39807, OSTA, OSTalphaApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-797