Learn More
ACROBiosystems Recombinant Protein;Human Osteoactivin / GPNMB Protein;HEK293;1MG;GPB,GPNMB,HGFIN,NMB,Osteoactivin,DC-HIL,OA
Catalog No. 501658373
Encompass_Preferred
Recombinant Protein;Human Osteoactivin / GPNMB Protein;1MG;Human GPNMB, His Tag (GPB-H5229) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Pro 486 (Accession # AAH32783).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 52.9 kDa. The protein migrates as 85-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8373 Supplier ACROBiosystems Supplier No. GPBH52291MG
Recombinant Protein;Human Osteoactivin / GPNMB Protein;1MG;Human GPNMB, His Tag (GPB-H5229) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Pro 486 (Accession # AAH32783).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 52.9 kDa. The protein migrates as 85-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.