Learn More
Promocell GmbH PromoCell Osteoblast Basal Medium (prf) 500 ml, phenol red free
Catalog No. 50306262
Encompass
PromoCell Osteoblast Growth Medium has been optimized for the in vitro cultivation of human osteoblasts. The fetal calf serum in the medium is strictly pretested for optimal performance.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for bovine, murine, and rat osteoblasts as well as osteoblast cell lines.Components:* Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMix* SupplementMix: Contains all media supplements premixed in one vial| Media supplement | Final supplement concentration|| Fetal Calf Serum | 0.1 ml / ml |
Catalog No. 50-306-262 Supplier Promocell GmbH Supplier No. C27015
PromoCell Osteoblast Growth Medium has been optimized for the in vitro cultivation of human osteoblasts. The fetal calf serum in the medium is strictly pretested for optimal performance.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for bovine, murine, and rat osteoblasts as well as osteoblast cell lines.Components:* Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMix* SupplementMix: Contains all media supplements premixed in one vial| Media supplement | Final supplement concentration|| Fetal Calf Serum | 0.1 ml / ml |