Learn More
Promocell GmbH PromoCell Osteoblast Mineralization Medium 100 ml, ready-to-use, with C-39616 attached to the flask
Catalog No. 50306263
Encompass
PromoCell Osteoblast Mineralization Medium has been developed for the efficient induction of strong mineralization in human osteoblast cultures. Extensive extracellular calcium-deposits can be observed after 21 days by following an easy one-step protocol.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for bovine, murine, and rat osteoblasts as well as osteoblast cell lines.To view a list of references where this medium was used with other species/cell types please check the reference literature tab on the manufacturerer homepage.
Catalog No. 50-306-263 Supplier Promocell GmbH Supplier No. C27020
PromoCell Osteoblast Mineralization Medium has been developed for the efficient induction of strong mineralization in human osteoblast cultures. Extensive extracellular calcium-deposits can be observed after 21 days by following an easy one-step protocol.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for bovine, murine, and rat osteoblasts as well as osteoblast cell lines.To view a list of references where this medium was used with other species/cell types please check the reference literature tab on the manufacturerer homepage.