Learn More
CPC Scientific H-Gly-Phe-Gln-Glu-Ala-Tyr-Arg-Arg-Phe-Tyr-Gly-Pro-Val-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific OSTP002B
Encompass
ONE-LETTER SEQUENCE: GFQEAYRRFYGPVMOLECULAR FORMULA: C75H104N20O19MOLECULAR WEIGHT:1589.77STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [89458-24-2], RESEARCH AREA: OsteoporosisREFERENCES: J.W. Poser et al., PNAS, 255, 8685 (1980)
Catalog No. 50-211-0002