Learn More
Cayman Chemical OsteocalcIn1-49humnt 100ug
SDP
Catalog No. 502936542
Encompass_Preferred
A bone peptide; secreted by osteoblasts and odontoblasts and comprises 1-2% of the total protein in bone; plasma levels increase in diseases that induce dysregulated bone turnover; positively correlated with insulin sensitivity and negatively correlated with high blood glucose levels in women; induces chemotaxis of MDA-MB-231 breast cancer cells, human peripheral blood monocytes, and rat osteosarcoma cells with osteoblast-like characteristics; positively associated with calcification of aortic tissue and heart valves in humans
Catalog No. 50-293-6542 Supplier Cayman Chemical Supplier No. 24723100 UG
A bone peptide; secreted by osteoblasts and odontoblasts and comprises 1-2% of the total protein in bone; plasma levels increase in diseases that induce dysregulated bone turnover; positively correlated with insulin sensitivity and negatively correlated with high blood glucose levels in women; induces chemotaxis of MDA-MB-231 breast cancer cells, human peripheral blood monocytes, and rat osteosarcoma cells with osteoblast-like characteristics; positively associated with calcification of aortic tissue and heart valves in humans