Learn More
STEMCELL Technologies MesenCult™ Osteogenic Differentiation Kit (Human), Size: 1 Kit
SDP
Catalog No. NC1598652
Encompass
MesenCult™ Osteogenic Differentiation Kit (Human) is specifically formulated for the in vitro differentiation of primary human mesenchymal stromal cells, and hPSC-derived mesenchymal progenitor cells (also known as mesenchymal stem cells or MSCs) into cells of the osteogenic lineage. This kit is suitable for the differentiation of human bone marrow (BM)- or adipose-derived MSCs previously culture-expanded in serum-containing medium (e.g. MesenCult™ Proliferation Kit [Human; Catalog #05411] or MesenCult™-hPL Medium [Human; Catalog #05439]) or animal component-free MesenCult™-ACF Plus Medium [Catalog #05445]).
This item is not returnable. View return policy
Catalog No. NC1598652 Supplier STEMCELL Technologies Supplier No. 05465
MesenCult™ Osteogenic Differentiation Kit (Human) is specifically formulated for the in vitro differentiation of primary human mesenchymal stromal cells, and hPSC-derived mesenchymal progenitor cells (also known as mesenchymal stem cells or MSCs) into cells of the osteogenic lineage. This kit is suitable for the differentiation of human bone marrow (BM)- or adipose-derived MSCs previously culture-expanded in serum-containing medium (e.g. MesenCult™ Proliferation Kit [Human; Catalog #05411] or MesenCult™-hPL Medium [Human; Catalog #05439]) or animal component-free MesenCult™-ACF Plus Medium [Catalog #05445]).