Learn More
Medchemexpress LLC Osteogenic growth peptide, ogp | 132996-61-3 | 99.2% | 1523.74 | 5 MG
SDP
Catalog No. 5000424330
Encompass_Preferred
Osteogenic Growth Peptide (OGP) is a naturally occurring 14-mer growth factor peptide found in serum. It is designed for research use, regulating proliferation, differentiation, and matrix mineralization in osteoblast lineage cells. Its active portion, OGP(10-14), binds to the OGP receptor, activating MAP kinase, Src, and RhoA pathways. OGP's primary structure is identical to the C-terminus of histone H4, and it is isolated from blood during osteogenic remodeling.
  • Naturally occurring 14-mer growth factor peptide
  • Regulates proliferation, differentiation, and matrix mineralization in osteoblast lineage cells
  • Active portion OGP(10-14) activates MAP kinase, Src, and RhoA pathways
  • Primary structure identical to the C-terminus of histone H4
  • Stimulates proliferation, differentiation, alkaline phosphatase activity, and matrix mineralization of osteoblastic lineage cells
  • For research use only
Catalog No. 50-004-24330 Supplier Medchemexpress LLC Supplier No. HYP15635MG
Osteogenic Growth Peptide (OGP) is a naturally occurring 14-mer growth factor peptide found in serum. It is designed for research use, regulating proliferation, differentiation, and matrix mineralization in osteoblast lineage cells. Its active portion, OGP(10-14), binds to the OGP receptor, activating MAP kinase, Src, and RhoA pathways. OGP's primary structure is identical to the C-terminus of histone H4, and it is isolated from blood during osteogenic remodeling.
  • Naturally occurring 14-mer growth factor peptide
  • Regulates proliferation, differentiation, and matrix mineralization in osteoblast lineage cells
  • Active portion OGP(10-14) activates MAP kinase, Src, and RhoA pathways
  • Primary structure identical to the C-terminus of histone H4
  • Stimulates proliferation, differentiation, alkaline phosphatase activity, and matrix mineralization of osteoblastic lineage cells
  • For research use only