Learn More
Osteometric Board
SDPSCI_ED
Catalog No. s05971
Change view
Click to view available options
:
70 x 13 11cm; 1mm increments
Catalog No.
S05971 70 x 13 11cm; 1mm increments
1 options
Catalog No. S05971 Supplier Science First Supplier No. 6346100
Only null left
Add to Cart
Add to Cart
Includes
Instructions
Useful measuring tool for every forensic lab.
  • Designed for making linear measurements of long bones, such as the femur or humerus, that are too long for calipers
  • Linear scale (60cm in 1mm increments) is screen printed directly onto base of measuring surface for reliable, high-precision results
  • Constructed of lightweight styrene for durability
  • Dimensions: 27.5 × 5.1 × 4.3 in. (70 × 13 × 11cm)