Learn More
Med Vet International Osteophyte Rasp, J0209R, Each
Catalog No. 5000585856
Encompass_Preferred
Most stifles requiring cruciate repair will exhibit some osteophytes. In some cases they may become very bulky necessitating removal. The osteophyte rasp is a small yet very effective rasp to keep in the cruciate set.
Catalog No. 50-005-85856 Supplier Med Vet International Supplier No. JOR209R
Most stifles requiring cruciate repair will exhibit some osteophytes. In some cases they may become very bulky necessitating removal. The osteophyte rasp is a small yet very effective rasp to keep in the cruciate set.