Learn More
Biomatik Corporation Osteopontin Polyclonal Antibody
Supplier:  Biomatik Corporation CAF50243
Encompass_Preferred
Osteopontin Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Secreted Phosphoprotein 1. The Osteopontin Polyclonal Antibody has been validated for the following applications: ELISA, IHC, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Please Inquire. Applications: ELISA, IHC, WB, IF. Target Synonyms: Secreted Phosphoprotein 1. Accession Number: P10451. Gene ID: 6696. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9224