Learn More
Sigma Aldrich Fine Chemicals Biosciences Osteopontin from bovine mi
Catalog No. 502904676
Encompass
Osteopontin (OPN) is a multifunctional protein. It is released by macrophages T cells and epithelial cells. It has 261 amino acid residues with a molecular mass of 29 283nbspDa. Osteopontin is also called as uropontin. Osteopontin from bovine milk is highly glycosylated and phosphorylated. SPP1 gene (secreted phosphoprotein 1) codes for osteopontin. This gene is located on human chromosome 4 and cow chromosome 6. It has seven exons and six introns.
Catalog No. 50-290-4676 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. O351450UG
Osteopontin (OPN) is a multifunctional protein. It is released by macrophages T cells and epithelial cells. It has 261 amino acid residues with a molecular mass of 29 283nbspDa. Osteopontin is also called as uropontin. Osteopontin from bovine milk is highly glycosylated and phosphorylated. SPP1 gene (secreted phosphoprotein 1) codes for osteopontin. This gene is located on human chromosome 4 and cow chromosome 6. It has seven exons and six introns.