Learn More
BioVendor, LLC Osteopontin Human Elisa BioVendor LLC
SDP
Supplier:  BioVendor, LLC RBL009R
Encompass
Osteopontin (OPN), a secreted multifunctional phosphoprotein, is a member of the small integrin-binding ligand N-linked glycoprotein (SIBLING) family of cell matrix proteins and participates in many biological activities. OPN has been demonstrated to be closely related to the occurrence and development of many bone-related diseases, such as osteoporosis, rheumatoid arthritis, and osteosarcoma. Besides that, it functions as a proinflammatory cytokine and promotes cell-mediated immune responses, and also has protective functions such as biomineralization and wound healing, and is also a strong predictor of adverse outcomes in patients with CVDs. Thus, OPN is not only a risk factor but also a potential therapeutic target for CVDs. One kit (96 wells). Type: sandwich ELISA. Sample type: serum, plasma. Sample req: 6 ¿l/well. Cal range: 125-4000pg/ml. LoD: 31pg/ml. Other names: OPN.
Catalog No. 50-240-6366