Learn More
Enzo Life Sciences Osteopontin monoclonal antibody (53) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS7530100
Encompass_Preferred
Clone: 53. Isotype: IgG2a. Immunogen: Recombinant human Osteopontin. Source: Purified from hybridoma tissue culture supernatant. . Species reactivity: Human. Applications: ELISA, IHC (FS), IHC (PS), WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2,containing 0.1% sodium azide.
Catalog No. 50-201-3292