Learn More
Enzo Life Sciences Osteopontin polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ALX210309R100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 59-74 (P59SKSNESHDHMDDMDD74) of mature human osteopontin. Species reactivity: Human, Mouse. Applications: ELISA, IHC (PS), WB. Formulation: Lyophilized. From neat/unpurified rabbit serum.
Catalog No. 50-200-8105