Learn More
ACROBiosystems Recombinant Protein;Human Osteopontin (OPN) Protein;HEK293;1MG;OPN,SPP1,BNSP,OPN,Uropontin,Nephropontin,Osteopontin,SPP-1,BSP-1,ETA-1,BSPI,BSP1,ETA1
Supplier:  ACROBiosystems OPNH52271MG
Encompass_Preferred
Recombinant Protein;Human Osteopontin (OPN) Protein;1MG;Human OPN, His Tag (OPN-H5227) is expressed from human 293 cells (HEK293). It contains AA Ile 17 - Asn 300 (Accession # AAH07016).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 33 kDa. The protein migrates as 60-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8040