Learn More
Alpco Ltd Osteoprotegerin ELISA (OPG ELISA)
SDP
Supplier:  Alpco Ltd 04BI20403
Encompass
Supplied with two kit controls, CTRL A and CTRL B, the Osteoprotegerin ELISA (OPG ELISA) is intended for the quantitative determination of osteoprotegerin in human plasma, serum or cell culture supernatants.
  • Includes ready to use controls and standards
  • Broad Detection Range: 25-400 pg/mL
  • Colorimetric Detection
  • Excellent precision and reproducibility:
    • Intra-assay CV 2-3%
    • Inter-assay CV 3-5%
 For Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-305-073