Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Osteoprotegerin (TNFRSF11B) Instant ELISA™ Kit

Catalog No. 5017898
Encompass_Preferred
Change view
Click to view available options
Quantity:
128 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-178-98 128 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-178-98 Supplier Invitrogen™ Supplier No. BMS2021INST
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Human Osteoprotegerin (TNFRSF11B) (Hu OPG) ELISA quantitates Hu OPG in human serum, plasma, cell culture supernatants or other body fluids. The assay will exclusively recognize both natural and recombinant Hu OPG. Principle of the method The Human OPG solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The protein encoded by this gene is a member of the TNF-receptor superfamily. This protein is an osteoblast-secreted decoy receptor that functions as a negative regulator of bone resorption. This protein specifically binds to its ligand, osteoprotegerin ligand, both of which are key extracellular regulators of osteoclast development. Studies of the mouse counterpart also suggest that this protein and its ligand play a role in lymph-node organogenesis and vascular calcification. Alternatively spliced transcript variants of this gene have been reported, but their full length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O00300
Assay Range 15.6-1000 pg/mL
Assay Sensitivity 2.5 pg/mL
Content And Storage Aluminum pouch(es) with a Microplate coated with Polyclonal Antibody to OPG, Biotin-Conjugate (anti-OPG polyclonal antibody), Streptavidin-HRP and Sample Diluent, lyophilized
Aluminum pouch(es) with a human Osteoprotegerin Standard curve (coloured)
Wash Buffer Concentrate 20x (phosphate-buffered saline with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Sample Diluent (Use when an external predilution of the samples is needed)
Stop Solution (1M Phosphoric acid)
Adhesive Plate Covers
Conjugate HRP
Immunoassay Kit Format Coated ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Application) ELISA
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias LOW QUALITY PROTEIN: tumor necrosis factor receptor superfamily member 11B; MGC29565; OCIF; Opg; osteoclastogenesis inhibitory factor; Osteoprotegerin; PDB5; soluble OPG; soluble Osteoprotegerin; sOPG; TNF receptor superfamily member 11b; TNFRSF11B; TR1; tumor necrosis factor receptor superfamily member 11B; tumor necrosis factor receptor superfamily, member 11b; tumor necrosis factor receptor superfamily, member 11b (osteoprotegerin)LOW QUALITY PROTEIN: tumor necrosis factor receptor superfamily member 11B; MGC29565; OCIF; Opg; osteoclastogenesis inhibitory factor; Osteoprotegerin; PDB5; soluble OPG; soluble Osteoprotegerin; sOPG; TNF receptor superfamily member 11b; TNFRSF11B; TR1; tumor necrosis factor receptor superfamily member 11B; tumor necrosis factor receptor superfamily, member 11b; tumor necrosis factor receptor superfamily, member 11b (osteoprotegerin); OCIF; OPG; PDB5; TR1
Gene ID (Entrez) 4982
Gene Symbol TNFRSF11B
Interassay CV 8%
Intraassay CV 7%
Kit Contents Pre-coated 96 well Microplate, Biotinylated Detection Antibody, SAV-HRP, Sample Diluent, lyophilized, Standard (8-well strips x 4), Wash Buffer Concentrate, Substrate Solution, Sample Diluent, Stop Solution, Lysis Buffer 10x, Adhesive Plate Covers, Biotinylated Detection Antibody, SAV-HRP, Sample Diluent, lyophilized, Standard (8-well strips x 4), Wash Buffer Concentrate, Substrate Solution, Sample Diluent, Stop Solution, Lysis Buffer 10x, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases Osteoclastogenesis inhibitory factor, Osteoprotegerin
Quantity 128 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements -20°C
Synonym Osteoprotegerin, TNF-R superfamily member 11 b
Target TNFRSF11B
Test Time 15 min
Total Assay Time 3 hr 10 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.