Learn More
ACROBiosystems Recombinant Protein;Human Osteoprotegerin / TNFRSF11B Protein, Fc Tag;HEK293;1MG;TNB,TNFRSF11B,OCIF,OPG,Osteoprotegerin
Supplier:  ACROBiosystems TNBH52591MG
Encompass_Preferred
Recombinant Protein;Human Osteoprotegerin / TNFRSF11B Protein, Fc Tag;1MG;Human OPG, Fc Tag (TNB-H5259) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Leu 401 (Accession # NP_002537).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 70.2 kDa. As a result of glycosylation, the protein migrates as 66-85 kDa under reducing (R) condition, and 130-170 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-165-8378