Learn More
ACROBiosystems Recombinant Protein;Human Osteoprotegerin / TNFRSF11B Protein;HEK293;1MG;TNB,TNFRSF11B,OCIF,OPG,Osteoprotegerin
Catalog No. 501658380
Encompass_Preferred
Recombinant Protein;Human Osteoprotegerin / TNFRSF11B Protein;1MG;Human OPG, His Tag (TNB-H5220) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Leu 401 (Accession # NP_002537).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 44.4 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TNFSF11, Fc tag (Cat. No. RAL-H5265) at 10 μg/mL (100 μL/well) can bind Human OPG, His Tag (Cat. No. TNB-H5220) with a linear range of 5-156 ng/mL (QC tested).
Catalog No. 50-165-8380 Supplier ACROBiosystems Supplier No. TNBH52201MG
Recombinant Protein;Human Osteoprotegerin / TNFRSF11B Protein;1MG;Human OPG, His Tag (TNB-H5220) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Leu 401 (Accession # NP_002537).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 44.4 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TNFSF11, Fc tag (Cat. No. RAL-H5265) at 10 μg/mL (100 μL/well) can bind Human OPG, His Tag (Cat. No. TNB-H5220) with a linear range of 5-156 ng/mL (QC tested).