Learn More
CPC Scientific H-Thr-Arg-Ser-Ala-Trp-Leu-Asp-Ser-Gly-Val-Thr-Gly-Ser-Gly-Leu-Glu-Gly-Asp-His-Leu-Ser-Asp-Thr-Ser-Thr-Thr-Ser-Leu-Glu-Leu-Asp-Ser-Arg-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110011
Encompass
This peptide is the carboxyl-terminal fragment of pTHrP, which is a potent inhibitor of osteoclastic bone resorption. It is suggested that this product acts directly on osteoclasts.ONE-LETTER SEQUENCE: TRSAWLDSGVTGSGLEGDHLSDTSTTSLELDSRMOLECULAR FORMULA: C142H228N42O58MOLECULAR WEIGHT:3451.62STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [137348-10-8], RESEARCH AREA: OsteoporosisREFERENCES: A.J.Fenton et al., Endocrinology, 129, 1762 (1991); A.J.Fenton et al., Endocrinology, 129, 3424 (1991)
Catalog No. 50-211-0011 Supplier CPC Scientific Supplier No. OSTP008A
This peptide is the carboxyl-terminal fragment of pTHrP, which is a potent inhibitor of osteoclastic bone resorption. It is suggested that this product acts directly on osteoclasts.ONE-LETTER SEQUENCE: TRSAWLDSGVTGSGLEGDHLSDTSTTSLELDSRMOLECULAR FORMULA: C142H228N42O58MOLECULAR WEIGHT:3451.62STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [137348-10-8], RESEARCH AREA: OsteoporosisREFERENCES: A.J.Fenton et al., Endocrinology, 129, 1762 (1991); A.J.Fenton et al., Endocrinology, 129, 3424 (1991)