Learn More
CPC Scientific H-Thr-Arg-Ser-Ala-Trp-Leu-Asp-Ser-Gly-Val-Thr-Gly-Ser-Gly-Leu-Glu-Gly-Asp-His-Leu-Ser-Asp-Thr-Ser-Thr-Thr-Ser-Leu-Glu-Leu-Asp-Ser-Arg-NH2
(trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific OSTP009A
Encompass
pTHrP (107-139) stimulates IL-6 expression in human osteoblastic cells. It inhibits bone resorption in vivoONE-LETTER SEQUENCE: TRSAWLDSGVTGSGLEGDHLSDTSTTSLELDSR-NH2MOLECULAR FORMULA: C142H229N43O57MOLECULAR WEIGHT:3450.64STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [204383-55-1], RESEARCH AREA: OsteoporosisREFERENCES: A.J.Fenton et al., Endocrinology, 129, 1762 (1991); A.J.Fenton et al., Endocrinology, 129, 3424 (1991); J.Cornish et al., Endocrinology, 138, 1299 (1997); F.de Miguel et al., J. Am. Soc. Nephrol., 10, 796 (1999); A.Valn et al., Endocrinology, 142, 2752 (2001)
Catalog No. 50-211-0013