Learn More
Bachem Osteostatin (human) Trifluoroacetate
SDP
Supplier:  Bachem 4031194.0500
Encompass_Preferred
Osteostatin (human) Trifluoroacetate, H-Thr-Arg-Ser-Ala-Trp-Leu-Asp-Ser-Gly-Val-Thr-Gly-Ser-Gly-Leu-Glu-Gly-Asp-His-Leu-Ser-Asp-Thr-Ser-Thr-Thr-Ser-Leu-Glu-Leu-Asp-Ser-Arg-OH, 137348-10-8, C142H228N42O58, Mr 3451.62, 0.5mg. Hypercalcemia of Malignancy Factor (107-139) (human), pTH-Related Protein (107-139) (human). The carboxyl-terminal fragment of pTHrP, TRSAWLDSGVTGSGLEGDHLSDTSTTSLELDSR, is a potent inhibitor of osteoclastic bone resorption. It is suggested that this product acts directly on osteoclasts.
Catalog No. 50-259-722