Learn More
World Precision Instrument Osteotomes- Bone Chisel, Lambotte, Straight, 2.5cm
SDP
Catalog No. 502538376
Encompass
Orthopedic surgeons us the straight Lambotte Osteotomes, bone chisels, to shape hard and dense bones. These are typically used when repairing fractured bones or when repairing a joint, whether from injury or deformity. This chisel is crafted from standard grade stainless steel, suitable for all forms of disinfection and autoclaving. These are generally mallet-driven, and you can choose the width of the chiseled end, either 2.5 cm (1") or 1.3 cm (0.5") wide.
Catalog No. 50-253-8376 Supplier World Precision Instrument Supplier No. WP006C
Orthopedic surgeons us the straight Lambotte Osteotomes, bone chisels, to shape hard and dense bones. These are typically used when repairing fractured bones or when repairing a joint, whether from injury or deformity. This chisel is crafted from standard grade stainless steel, suitable for all forms of disinfection and autoclaving. These are generally mallet-driven, and you can choose the width of the chiseled end, either 2.5 cm (1") or 1.3 cm (0.5") wide.