Learn More
World Precision Instrument Osteotomes - Bone Chisel, U.S. Army Pattern, 18mm
SDP
Catalog No. 502538358
Encompass
This Osteotome, bone chisel, can be used for scraping, cleaning and sculpting bone. For example, you could chisel out bone fragments to access restricted spaces. Three tip sizes are available (6, 12, or 18mm), one for every application. This chisel is crafted from standard grade stainless steel, suitable for all forms of disinfection and autoclaving.
Catalog No. 50-253-8358 Supplier World Precision Instrument Supplier No. WP0419
This Osteotome, bone chisel, can be used for scraping, cleaning and sculpting bone. For example, you could chisel out bone fragments to access restricted spaces. Three tip sizes are available (6, 12, or 18mm), one for every application. This chisel is crafted from standard grade stainless steel, suitable for all forms of disinfection and autoclaving.