Learn More
Medchemexpress LLC HY-N2554 1mg Medchemexpress, Osthenol CAS:484-14-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN25541MG
Encompass_Preferred
Medchemexpress, HY-N2554 1mg Osthenol CAS:484-14-0 Osthenol (Ostenol), a prenylated coumarin isolated from the dried roots of Angelica pubescens, is selective, reversible, and competitive human monoamine oxidase-A (hMAO-A) inhibitor (Ki=0.26 µM). Osthenol potently inhibits recombinant hMAO-A with an IC50 of 0.74 µM and shows a high selectivity index for hMAO-A versus hMAO-B. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2902