Learn More
Medchemexpress LLC HY-N0054 250mg Medchemexpress, Osthole CAS:484-12-8 Purity:>98%
SDP
Catalog No. 501933373
Encompass_Preferred
Medchemexpress, HY-N0054 250mg Osthole CAS:484-12-8 Osthole is a natural antihistamine alternative. Osthole may be a potential inhibitor of histamine H 1 receptor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3373 Supplier Medchemexpress LLC Supplier No. HYN0054250MG
Medchemexpress, HY-N0054 250mg Osthole CAS:484-12-8 Osthole is a natural antihistamine alternative. Osthole may be a potential inhibitor of histamine H 1 receptor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.