Learn More
Med Vet International Ostifen (Carprofen) Caplets,Dogs,100mg,60 Count
Catalog No. 502828186
Encompass_Preferred
Carprofen is indicated for the relief of pain and inflammation associated with osteoarthritis and for the control of postoperative pain associated with soft tissue and orthopedic surgeries in dogs. See package insert for complete product details. *Based on product availability, OstiFen (Carprofen) Caplets may be substituted with Carprieve (Carprofen) Caplets.
Catalog No. 50-282-8186 Supplier Med Vet International Supplier No. RXOSTIFEN10060
Carprofen is indicated for the relief of pain and inflammation associated with osteoarthritis and for the control of postoperative pain associated with soft tissue and orthopedic surgeries in dogs. See package insert for complete product details. *Based on product availability, OstiFen (Carprofen) Caplets may be substituted with Carprieve (Carprofen) Caplets.