Learn More
Med Vet International Ostifen (Carprofen) Chewable Tablets,Dogs,75mg,60 Count
Catalog No. 502828857
Encompass_Preferred
Carprofen is indicated for the relief of pain and inflammation associated with osteoarthritis and for the control of postoperative pain associated with soft tissue and orthopedic surgeries in dogs. See package insert for complete product details. *Based on product availability, OstiFen (Carprofen) Caplets may be substituted with Carprieve (Carprofen) Caplets.
Catalog No. 50-282-8857 Supplier Med Vet International Supplier No. RXOSTIFEN7560CH
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Carprofen is indicated for the relief of pain and inflammation associated with osteoarthritis and for the control of postoperative pain associated with soft tissue and orthopedic surgeries in dogs. See package insert for complete product details. *Based on product availability, OstiFen (Carprofen) Caplets may be substituted with Carprieve (Carprofen) Caplets.