Learn More
Med Vet International Ostimax (Deracoxib) Chewable Tablets For Dogs, 12mg, 90 Count
Catalog No. 502828606
Encompass_Preferred
OstiMax Chewable Tablets (deracoxib) are indicated for the control of post-operative pain and inflammation associated with orthopedic (bone) and dental surgery in dogs and for the control of pain and inflammation (soreness) associated with osteoarthritis in dogs.
Catalog No. 50-282-8606 Supplier Med Vet International Supplier No. RXOSTI1290V
OstiMax Chewable Tablets (deracoxib) are indicated for the control of post-operative pain and inflammation associated with orthopedic (bone) and dental surgery in dogs and for the control of pain and inflammation (soreness) associated with osteoarthritis in dogs.