Learn More
Sino Biological OSTM1 Antibody, Rabbit PAb 100uL
Supplier:  Sino Biological 10913RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human OSTM1 extracellular domain (rh OSTM1; Catalog#10913-H08H; NP_054747.2; Met 1-Pro 284). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-224-7937