Learn More
Sigma Aldrich Fine Chemicals Biosciences Ostwald viscometer tube ou1EA
Catalog No. 502904735
Encompass
for the determination of relative viscosities of liquids. Adjusted for outflow time of 80 to 100s with water at 20C.
Catalog No. 50-290-4735 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2754091EA
for the determination of relative viscosities of liquids. Adjusted for outflow time of 80 to 100s with water at 20C.