Learn More
Selleck Chemical LLC OSU-03012 (AR-12) 25mg 742112-33-0
SDP
Catalog No. 508857
Encompass_Preferred
OSU-03012 (AR-12) is a potent inhibitor of recombinant PDK-1(phosphoinositide-dependent kinase 1) with IC50 of 5 ?M in a cell-free assay and 2-fold increase in potency over OSU-02067. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-885-7 Supplier Selleck Chemical LLC Supplier No. S110625MG
OSU-03012 (AR-12) is a potent inhibitor of recombinant PDK-1(phosphoinositide-dependent kinase 1) with IC50 of 5 ?M in a cell-free assay and 2-fold increase in potency over OSU-02067. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.