Learn More
TARGETMOL CHEMICALS INC OSU 6162 5MG
SDP
Catalog No. 502579697
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (+)-OSU 6162 (Piperidine 3-[3-(methylsulfonyl)phenyl]-1-propyl- (3R)-) is an agonist of 5-HT Receptor with anti-Alzheimer and antidepressant activities. purity: 99%
Catalog No. 50-257-9697 Supplier TARGETMOL CHEMICALS INC Supplier No. T600275MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (+)-OSU 6162 (Piperidine 3-[3-(methylsulfonyl)phenyl]-1-propyl- (3R)-) is an agonist of 5-HT Receptor with anti-Alzheimer and antidepressant activities. purity: 99%