Learn More
Selleck Chemical LLC OSU-T315
SDP
Catalog No. 5000002711
Encompass_Preferred
OSU-T315 (ILK-IN-2) is a potent Integrin-Linked Kinase (ILK) inhibitor with IC50 of 0 6 M OSU-T315 induces autophagy and apoptosis both of which are integral to its antiproliferative activity OSU-T315 exhibits anti-tumor activity
Catalog No. 50-000-02711 Supplier Selleck Chemical LLC Supplier No. S68405MG
OSU-T315 (ILK-IN-2) is a potent Integrin-Linked Kinase (ILK) inhibitor with IC50 of 0 6 M OSU-T315 induces autophagy and apoptosis both of which are integral to its antiproliferative activity OSU-T315 exhibits anti-tumor activity