Learn More
TARGETMOL CHEMICALS INC OSU03012 25MG
SDP
Catalog No. 502580790
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Osu03012 (AR-12) is an orally bioavailable small-molecule celecoxib-derived inhibitor of phosphoinositide-dependent kinase-1 (PDK1) with potential antineoplastic activity. purity: 99%
Catalog No. 50-258-0790 Supplier TARGETMOL CHEMICALS INC Supplier No. T246625MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Osu03012 (AR-12) is an orally bioavailable small-molecule celecoxib-derived inhibitor of phosphoinositide-dependent kinase-1 (PDK1) with potential antineoplastic activity. purity: 99%